accessibility menu, dialog, popup

UserName

BrBr

CAS RN 75415-78-0

IUPAC Name: 1,2-dibromocyclopent-1-ene
Synonyms: 1,2-dibromocyclopent-1-ene
Molecular Weight (g/mol): 225.91
Molecular Formula: C5H6Br2
InChi Key: PNWFXPGGROADNS-UHFFFAOYSA-N
SMILES: BrC1=C(Br)CCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)