accessibility menu, dialog, popup

UserName

OSOOH

CAS RN 75434-70-7

IUPAC Name: 2-[(2-phenoxyethyl)sulfanyl]acetic acid
Synonyms: [(2-phenoxyethyl)sulfanyl]acetic acid
Molecular Weight (g/mol): 212.26
Molecular Formula: C10H12O3S
InChi Key: SHWKIHWUFNTMIB-UHFFFAOYSA-N
SMILES: OC(=O)CSCCOC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results