accessibility menu, dialog, popup

UserName

NH2SNN

CAS RN 7552-07-0

IUPAC Name: 1,2,4-thiadiazol-5-amine
Synonyms: 1,2,4-thiadiazol-5-amine
Molecular Weight (g/mol): 101.13
Molecular Formula: C2H3N3S
InChi Key: VJHTZTZXOKVQRN-UHFFFAOYSA-N
SMILES: NC1=NC=NS1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results