accessibility menu, dialog, popup

UserName

H3CONH2NCl

CAS RN 75711-01-2

IUPAC Name: 6-chloro-5-methoxypyridin-3-amine
Synonyms: 6-chloro-5-methoxypyridin-3-amine
Molecular Weight (g/mol): 158.59
Molecular Formula: C6H7ClN2O
InChi Key: FJKVNEBFHWMVGF-UHFFFAOYSA-N
SMILES: COC1=C(Cl)N=CC(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results