accessibility menu, dialog, popup

UserName

NH2NOO

CAS RN 7579-20-6

IUPAC Name: 3-aminopyridine-4-carboxylate
Synonyms: 3-aminopyridine-4-carboxylate
Molecular Weight (g/mol): 137.12
Molecular Formula: C6H5N2O2
InChi Key: FYEQKMAVRYRMBL-UHFFFAOYSA-M
SMILES: NC1=CN=CC=C1C([O-])=O
Percent Purity 
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results