accessibility menu, dialog, popup

UserName

CAS RN 76003-29-7

CH3H3CCH3OONNHO

CAS RN 76003-29-7

IUPAC Name: tert-butyl 3-oxopiperazine-1-carboxylate
Synonyms: tert-butyl 3-oxopiperazine-1-carboxylate
Molecular Weight (g/mol): 200.24
Molecular Formula: C9H16N2O3
InChi Key: FCMLWBBLOASUSO-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCNC(=O)C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)