accessibility menu, dialog, popup

UserName

CH3CH3H2C

CAS RN 763-29-1

IUPAC Name: 2-methylpent-1-ene
Synonyms: 2-methyl-1-pentene
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: WWUVJRULCWHUSA-UHFFFAOYSA-N
SMILES: CCCC(C)=C
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)