accessibility menu, dialog, popup

UserName

H3CCH3HNO

CAS RN 766-36-9

IUPAC Name: 3-ethyl-4-methyl-2,5-dihydro-1H-pyrrol-2-one
Synonyms: 3-ethyl-4-methyl-1,5-dihydropyrrol-2-one
Molecular Weight (g/mol): 125.17
Molecular Formula: C7H11NO
InChi Key: YCTNTSVMJWIYTQ-UHFFFAOYSA-N
SMILES: CCC1=C(C)CNC1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results