accessibility menu, dialog, popup

UserName

H2NNO

CAS RN 7663-77-6

IUPAC Name: 1-(3-aminopropyl)pyrrolidin-2-one
Synonyms: 1-(3-aminopropyl)pyrrolidin-2-one
Molecular Weight (g/mol): 142.2
Molecular Formula: C7H14N2O
InChi Key: HJORCZCMNWLHMB-UHFFFAOYSA-N
SMILES: NCCCN1CCCC1=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)