accessibility menu, dialog, popup

UserName

CAS RN 76655-99-7

CH3SONH2

CAS RN 76655-99-7

IUPAC Name: 3-methylthiophene-2-carboxamide
Synonyms: 3-methylthiophene-2-carboxamide
Molecular Weight (g/mol): 141.19
Molecular Formula: C6H7NOS
InChi Key: QZDTWJRYMXQXBX-UHFFFAOYSA-N
SMILES: CC1=C(SC=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results