accessibility menu, dialog, popup

UserName

CH3OH3CCH3O

CAS RN 768-58-1

IUPAC Name: 2-ethyl-5,5-dimethyl-1,3-dioxane
Synonyms: 2-ethyl-5,5-dimethyl-1,3-dioxane
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: QSHOOPIYPOINNH-UHFFFAOYSA-N
SMILES: CCC1OCC(C)(C)CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results