accessibility menu, dialog, popup

UserName

CAS RN 7693-46-1

IUPAC Name: 4-nitrophenyl carbonochloridate
Synonyms: 4-nitrophenyl carbonochloridate
Molecular Weight (g/mol): 201.56
Molecular Formula: C7H4ClNO4
InChi Key: NXLNNXIXOYSCMB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OC(Cl)=O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (4)
Quantity 
  • (3)
  • (3)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (4)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (4)
  • (4)