accessibility menu, dialog, popup

UserName

CAS RN 77-49-6

IUPAC Name: 2-methyl-2-nitropropane-1,3-diol
Synonyms: 2-methyl-2-nitropropane-1,3-diol
Molecular Weight (g/mol): 135.12
Molecular Formula: C4H9NO4
InChi Key: LOTYADDQWWVBDJ-UHFFFAOYSA-N
SMILES: CC(CO)(CO)[N+]([O-])=O
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results