CAS RN 773-01-3
IUPAC Name:
2,4,6-trimethyl-1λ⁴-pyran-1-ylium; tetrafluoroboranuide
Synonyms:
2,4,6-trimethyl-1λ⁴-pyran-1-ylium tetrafluoroborate
Molecular Weight (g/mol):
209.98
Molecular Formula:
C8H11BF4O
InChi Key:
PYOPRIFMELCENN-UHFFFAOYSA-N
SMILES:
F[B-](F)(F)F.CC1=CC(C)=[O+]C(C)=C1
1
–
1
of
1
results
1
2,4,6-Trimethylpyrylium tetrafluoroborate, 98+%
CAS: 773-01-3 Molecular Formula: C8H11BF4O Molecular Weight (g/mol): 209.98 MDL Number: MFCD00032098 InChI Key: PYOPRIFMELCENN-UHFFFAOYSA-N Synonym: 2,4,6-trimethylpyrylium tetrafluoroborate,2,4,6-trimethyl-1??-pyran-1-ylium tetrafluoroborate,2,4,6-trimethylpyrylium tetrafluoborate,2,4,6-trimethylpyryliumtetrafluoroborate,pyrylium, 2,4,6-trimethyl-, tetrafluoroborate 1-,pyrylium, 2,4,6-trimethyl-, tetrafluoroborate 1-1:1,2,4,6-trimethyl-1$l^ 4-pyran-1-ylium tetrafluoroborate PubChem CID: 164732 IUPAC Name: 2,4,6-trimethylpyrylium;tetrafluoroborate SMILES: F[B-](F)(F)F.CC1=CC(C)=[O+]C(C)=C1