accessibility menu, dialog, popup

UserName

CAS RN 775-56-4

IUPAC Name: diethoxy(methyl)phenylsilane
Synonyms: silane, diethoxymethylphenyl-
Molecular Weight (g/mol): 210.35
Molecular Formula: C11H18O2Si
InChi Key: MNFGEHQPOWJJBH-UHFFFAOYSA-N
SMILES: CCO[Si](C)(OCC)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results