CAS RN 77500-04-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Amino-3,8-dimethylimidazo[4,5-f]quinoxaline, TRC
CAS: 77500-04-0 Molecular Formula: C11 H11 N5 Molecular Weight (g/mol): 213.24 Synonym: 3H-Imidazo[4,5-f]quinoxalin-2-amine, 3,8-dimethyl-,3,8-Dimethyl-3H-imidazo[4,5-f]quinoxalin-2-amine,2-Amino-3,8-dimethylimidazo[4,5-f]quinoxaline,2-Amino-3,8-dimethylmidazo[4,5-f]quinoxaline,MeIQx IUPAC Name: 3,8-dimethylimidazo[4,5-f]quinoxalin-2-amine SMILES: Cc1cnc2ccc3c(nc(N)n3C)c2n1