CAS RN 776-47-6
CAS RN 776-47-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Ethyl-5-fluoroindoline-2,3-dione, TRC
CAS: 776-47-6 Molecular Formula: C10H8NO2F Molecular Weight (g/mol): 193.17 Synonym: 1-Ethyl-5-fluoroindole-2,3-dione,1-Ethyl-5-fluoro-2,3-dihydro-1H-indole-2,3-dione,1-Ethyl-5-fluoro-1H-indole-2,3-dione,Isatin, 1-ethyl-5-fluoro- (6CI),Indole-2,3-dione, 1-ethyl-5-fluoro- (7CI,8CI) SMILES: CCN1C(=O)C(=O)c2cc(F)ccc12