accessibility menu, dialog, popup

UserName

OHMoOHOO

CAS RN 7782-91-4

IUPAC Name: dioxomolybdenumdiol
Synonyms: molybdic acid molybdic(VI) acid H2MoO4
Molecular Weight (g/mol): 161.96
Molecular Formula: H2MoO4
InChi Key: VLAPMBHFAWRUQP-UHFFFAOYSA-L
SMILES: O[Mo](O)(=O)=O
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (5)
Grade 
  • (2)
  • (5)
  • (2)
Physical Form 
  • (2)
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)