CAS RN 7783-51-9
IUPAC Name:
gallium(3+) trifluoride
Synonyms:
gallium(III) fluoride
Molecular Weight (g/mol):
126.72
Molecular Formula:
F3Ga
InChi Key:
WXXZSFJVAMRMPV-UHFFFAOYSA-K
SMILES:
[F-].[F-].[F-].[Ga+3]
1
–
2
of
2
results
1
Gallium(III) fluoride, anhydrous, 99.85% (metals basis), Thermo Scientific Chemicals
CAS: 7783-51-9 MDL Number: MFCD00011021
2
Gallium(III) fluoride, 99.995% (metals basis)
CAS: 7783-51-9 MDL Number: MFCD00011021