accessibility menu, dialog, popup

UserName

CAS RN 7783-86-0

FeII

CAS RN 7783-86-0

IUPAC Name: λ²-iron(2+) diiodide
Synonyms: iron(II) iodide
Molecular Weight (g/mol): 309.65
Molecular Formula: FeI2
InChi Key: BQZGVMWPHXIKEQ-UHFFFAOYSA-L
SMILES: [Fe++].[I-].[I-]
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)