CAS RN 77872-43-6
CAS RN 77872-43-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Hydroxy-N-isopropyl-N-methyltryptamine, TRC
CAS: 77872-43-6 Molecular Formula: C14 H20 N2 O Molecular Weight (g/mol): 232.32 Synonym: 1H-Indol-4-ol, 3-[2-[methyl(1-methylethyl)amino]ethyl]-,3-[2-[Methyl(1-methylethyl)amino]ethyl]-1H-indol-4-ol,4-Hydroxy-N-isopropyl-N-methyltryptamine,4-HO-MiPT,4-Hydroxy-N-methyl-N-isopropyltryptamine IUPAC Name: 3-[2-[methyl(propan-2-yl)amino]ethyl]-1H-indol-4-ol SMILES: CC(C)N(C)CCc1c[nH]c2cccc(O)c12