accessibility menu, dialog, popup

UserName

CAS RN 77876-39-2

IUPAC Name: [4-(diphenylphosphanyl)pentan-2-yl]diphenylphosphane
Synonyms: [4-(diphenylphosphanyl)pentan-2-yl]diphenylphosphane
Molecular Weight (g/mol): 440.51
Molecular Formula: C29H30P2
InChi Key: CTYPJIUQROQJBG-UHFFFAOYNA-N
SMILES: CC(CC(C)P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)