accessibility menu, dialog, popup

UserName

CAS RN 7789-59-5

BrPBrBrO

CAS RN 7789-59-5

IUPAC Name: phosphoroyl tribromide
Synonyms: phosphoryl bromide
Molecular Weight (g/mol): 286.69
Molecular Formula: Br3OP
InChi Key: UXCDUFKZSUBXGM-UHFFFAOYSA-N
SMILES: BrP(Br)(Br)=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (3)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)