accessibility menu, dialog, popup

UserName

CAS RN 7789-67-5

BrHSnBrHBrHHBr

CAS RN 7789-67-5

IUPAC Name: tin(IV) bromide
Synonyms: tin(IV) bromide
Molecular Weight (g/mol): 442.36
Molecular Formula: Br4H4Sn
InChi Key: QENMVORAJVNRHX-UHFFFAOYSA-N
SMILES: Br.Br.Br.Br.[Sn]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results