accessibility menu, dialog, popup

UserName

CAS RN 7790-45-6

IIIIIr

CAS RN 7790-45-6

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 699.84
Molecular Formula: I4Ir
InChi Key: UXDVYGYWEFPPTQ-UHFFFAOYSA-N
SMILES: [I].[I].[I].[I].[Ir]
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results