CAS RN 7796-19-2
CAS RN 7796-19-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Heptacosanone, TRC
CAS: 7796-19-2 Molecular Formula: C27H54O Molecular Weight (g/mol): 394.72 Synonym: Heptacosan-2-one IUPAC Name: heptacosan-2-one SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)C