accessibility menu, dialog, popup

UserName

OHNOO

CAS RN 7797-81-1

IUPAC Name: 3-hydroxy-3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Synonyms: 3-hydroxy-3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Molecular Weight (g/mol): 213.19
Molecular Formula: C12H7NO3
InChi Key: KTWCUGUUDHJVIH-UHFFFAOYSA-N
SMILES: ON1C(=O)C2=CC=CC3=CC=CC(C1=O)=C23
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results