accessibility menu, dialog, popup

UserName

CAS RN 78-90-0

CH3H2NH2N

CAS RN 78-90-0

IUPAC Name: propane-1,2-diamine
Synonyms: propylenediamine
Molecular Weight (g/mol): 74.13
Molecular Formula: C3H10N2
InChi Key: AOHJOMMDDJHIJH-UHFFFAOYNA-N
SMILES: CC(N)CN
Boiling Point 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)