accessibility menu, dialog, popup

UserName

NHNNH2Cl

CAS RN 78583-81-0

IUPAC Name: 5-(4-chlorophenyl)-1H-pyrazol-3-amine
Synonyms: 5-(4-chlorophenyl)-1H-pyrazol-3-amine
Molecular Weight (g/mol): 193.63
Molecular Formula: C9H8ClN3
InChi Key: XQPBZIITFQHIDI-UHFFFAOYSA-N
SMILES: NC1=NNC(=C1)C1=CC=C(Cl)C=C1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results