accessibility menu, dialog, popup

UserName

ONHNOO

CAS RN 78818-15-2

IUPAC Name: benzyl 3-oxopiperazine-1-carboxylate
Synonyms: benzyl 3-oxopiperazine-1-carboxylate
Molecular Weight (g/mol): 234.26
Molecular Formula: C12H14N2O3
InChi Key: BAHFPJFBMJTOPU-UHFFFAOYSA-N
SMILES: O=C(OCC1=CC=CC=C1)N1CCNC(=O)C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results