accessibility menu, dialog, popup

UserName

CH3CH3PHNPH3CCH3CH3H3CCH3H3CClIr

CAS RN 791629-96-4

IUPAC Name: bis({2-[bis(propan-2-yl)phosphanyl]ethyl})amine; chlorodihydridoiridium
Synonyms: bis[2-(diisopropylphosphanyl)ethyl]amine; chlorodihydridoiridium
Molecular Weight (g/mol): 535.11
Molecular Formula: C16H39ClIrNP2
InChi Key: QPAONQONOKDMMJ-UHFFFAOYSA-M
SMILES: Cl[IrH2].CC(C)P(CCNCCP(C(C)C)C(C)C)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results