CAS RN 79513-35-2
CAS RN 79513-35-2
IUPAC Name:
hydrogen tert-butyl N-(2-aminoethyl)carbamate chloride
Synonyms:
hydrogen tert-butyl N-(2-aminoethyl)carbamate chloride
Molecular Weight (g/mol):
196.68
Molecular Formula:
C7H17ClN2O2
InChi Key:
XUHJJLCKXZTUJN-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CC(C)(C)OC(=O)NCCN
1
–
1
of
1
results
1
N-Boc-ethylenediamine hydrochloride, 98%
CAS: 79513-35-2 Molecular Formula: C7H17ClN2O2 Molecular Weight (g/mol): 196.68 MDL Number: MFCD01076129 InChI Key: XUHJJLCKXZTUJN-UHFFFAOYSA-N Synonym: n-boc-ethylenediamine hydrochloride,tert-butyl 2-aminoethyl carbamate hydrochloride,tert-butyl n-2-aminoethyl carbamate hydrochloride,n-boc-ethane-1,2-diaminehcl,n-boc-ethane-1,2-diamine hcl,n-tert-butoxycarbonylethylenediamine hydrochloride,n-tert-butoxycarbonyl-ethylenediamine hydrochloride,n-boc-ethylenediamine hcl,tert-butyl n-2-aminoethyl carbamate-hcl,n-boc-ethylenediaminehydrochloride PubChem CID: 187200 IUPAC Name: tert-butyl N-(2-aminoethyl)carbamate;hydrochloride SMILES: [H+].[Cl-].CC(C)(C)OC(=O)NCCN