accessibility menu, dialog, popup

UserName

CAS RN 79709-85-6

IUPAC Name: 4-ethyl-4'-(4-pentylcyclohexyl)-1,1'-biphenyl
Synonyms: 4-ethyl-4'-(4-pentylcyclohexyl)-1,1'-biphenyl
Molecular Weight (g/mol): 334.55
Molecular Formula: C25H34
InChi Key: JOLGXBQYTARJLD-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC=C(CC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results