accessibility menu, dialog, popup

UserName

CH3NNNCO

CAS RN 799283-97-9

IUPAC Name: 4-isocyanato-5-methyl-1-phenyl-1H-pyrazole
Synonyms: 4-isocyanato-5-methyl-1-phenylpyrazole
Molecular Weight (g/mol): 199.21
Molecular Formula: C11H9N3O
InChi Key: IIFVDOYRTLEGHQ-UHFFFAOYSA-N
SMILES: CC1=C(C=NN1C1=CC=CC=C1)N=C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results