accessibility menu, dialog, popup

UserName

CAS RN 80126-53-0

CH3(S)NH2OHO

CAS RN 80126-53-0

IUPAC Name: (2S)-2-amino-3-(2-methylphenyl)propanoic acid
Synonyms: (2S)-2-amino-3-(2-methylphenyl)propanoic acid
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: NHBKDLSKDKUGSB-VIFPVBQESA-N
SMILES: CC1=CC=CC=C1C[C@H](N)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)