accessibility menu, dialog, popup

UserName

H3CSiH3CCH3OOONOO

CAS RN 80149-80-0

IUPAC Name: 4-nitrophenyl 2-(trimethylsilyl)ethyl carbonate
Synonyms: 4-nitrophenyl 2-(trimethylsilyl)ethyl carbonate
Molecular Weight (g/mol): 283.36
Molecular Formula: C12H17NO5Si
InChi Key: ZAQWGGKIMQIVGM-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CCOC(=O)OC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results