accessibility menu, dialog, popup

UserName

NCH3H2NNH2NNH2H2NClHClHCl

CAS RN 8063-24-9

IUPAC Name: 3,6-diamino-9-methylacridin-10-ium acridine-3,6-diamine chloride
Synonyms: 3,6-diamino-9-methylacridin-10-ium proflavine chloride
Molecular Weight (g/mol): 468.99
Molecular Formula: C27H25ClN6
InChi Key: PCVUBUSUCITCLW-UHFFFAOYSA-N
SMILES: [Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.CC1=C2C=CC(N)=CC2=[NH+]C2=CC(N)=CC=C12
Quantity 
  • (1)
  • (1)
  • (2)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results