CAS RN 80673-00-3
IUPAC Name:
3-[2-(trimethylsilyl)ethynyl]pyridine
Synonyms:
3-[2-(trimethylsilyl)ethynyl]pyridine
Molecular Weight (g/mol):
175.31
Molecular Formula:
C10H13NSi
InChi Key:
MTMBLPOPETZZRX-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)C#CC1=CN=CC=C1
1
–
1
of
1
results
1
3-[(Trimethylsilyl)ethynyl]pyridine, 97%
CAS: 80673-00-3 Molecular Formula: C10H13NSi Molecular Weight (g/mol): 175.306 MDL Number: MFCD01863744 InChI Key: MTMBLPOPETZZRX-UHFFFAOYSA-N Synonym: 3-trimethylsilyl ethynyl pyridine,3-trimethylsilylethynyl pyridine,3-pyridylethynyl trimethylsilane,3-2-trimethylsilyl ethynyl pyridine,trimethyl 2-pyridin-3-ylethynyl silane,acmc-209pjw,3-trimethylsilylethynylpyridine,trimethyl-2-3-pyridyl ethynyl silane,1-3-pyridyl-2-trimethylsilylacetylene PubChem CID: 4100857 IUPAC Name: trimethyl(2-pyridin-3-ylethynyl)silane SMILES: C[Si](C)(C)C#CC1=CN=CC=C1