CAS RN 80742-11-6
CAS RN 80742-11-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(2R,4R)-rel-1,2,3,4-Tetrahydro-6-methyl-2,4-bis(2-nitrophenyl)-5-pyrimidinecarboxylic Acid Methyl Ester, TRC
CAS: 80742-11-6 Molecular Formula: C19 H18 N4 O6 Molecular Weight (g/mol): 398.37 Synonym: 5-Pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-6-methyl-2,4-bis(2-nitrophenyl)-, methyl ester, (2R,4R)-rel-,5-Pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-6-methyl-2,4-bis(2-nitrophenyl)-, methyl ester, cis-(±)-,5-Pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-6-methyl-2,4-bis(2-nitrophenyl)-, methyl ester, cis-,Methyl (2RS,4RS)-6-Methyl-2,4-bis(2-nitrophenyl)-1,2,3,4-tetrahydropyrimidine-5-carboxylate IUPAC Name: methyl (2R,4R)-6-methyl-2,4-bis(2-nitrophenyl)-1,2,3,4-tetrahydropyrimidine-5-carboxylate SMILES: COC(=O)C1=C(C)N[C@@H](N[C@@H]1c2ccccc2[N+](=O)[O-])c3ccccc3[N+](=O)[O-]