accessibility menu, dialog, popup

UserName

CAS RN 80745-10-4

CH3SOOHNNOOHNH2NH2H3CH3COH3C(E/Z)

CAS RN 80745-10-4

IUPAC Name: 2-amino-5-{[amino(4-methoxy-2,3,6-trimethylbenzenesulfonamido)methylidene]amino}pentanoic acid
Synonyms: 2-amino-5-{[amino(4-methoxy-2,3,6-trimethylbenzenesulfonamido)methylidene]amino}pentanoic acid
Molecular Weight (g/mol): 386.47
Molecular Formula: C16H26N4O5S
InChi Key: FUSAEZSSVDNYPO-UHFFFAOYNA-N
SMILES: COC1=C(C)C(C)=C(C(C)=C1)S(=O)(=O)NC(N)=NCCCC(N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)