CAS RN 809-51-8
CAS RN 809-51-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7-Oxo Cholesterol 3-Acetate, TRC
CAS: 809-51-8 Molecular Formula: C29H46O3 Molecular Weight (g/mol): 442.68 Synonym: (3beta)-3-(Acetyloxy)cholest-5-en-7-one,3beta-Hydroxy-cholest-5-en-7-one Acetate,7-Ketocholesteryl Acetate,7-Oxocholest-5-en-3beta-yl Acetate,7-Oxocholesteryl Acetate,NSC 59469 IUPAC Name: [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-7-oxo-1,2,3,4,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-yl] acetate SMILES: CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@H]3[C@H](CC[C@]12C)[C@@]4(C)CC[C@@H](CC4=CC3=O)OC(=O)C