accessibility menu, dialog, popup

UserName

OO

CAS RN 81-21-0

IUPAC Name: 4,10-dioxapentacyclo[6.3.1.0²,⁷.0³,⁵.0⁹,¹¹]dodecane
Synonyms: dicyclopentadiene dioxide
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: BQQUFAMSJAKLNB-UHFFFAOYNA-N
SMILES: C1C2OC2C2C3CC(C4OC34)C12
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)