accessibility menu, dialog, popup

UserName

CH3OOSNCS

CAS RN 81321-10-0

IUPAC Name: methyl 3-isothiocyanatothiophene-2-carboxylate
Synonyms: methyl 3-isothiocyanatothiophene-2-carboxylate
Molecular Weight (g/mol): 199.24
Molecular Formula: C7H5NO2S2
InChi Key: RKBLURUDWDTMOB-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C=CS1)N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results