accessibility menu, dialog, popup

UserName

CH3OBr

CAS RN 816-40-0

IUPAC Name: 1-bromobutan-2-one
Synonyms: 1-bromo-2-butanone
Molecular Weight (g/mol): 151
Molecular Formula: C4H7BrO
InChi Key: CCXQVBSQUQCEEO-UHFFFAOYSA-N
SMILES: CCC(=O)CBr
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)