accessibility menu, dialog, popup

UserName

H3CCH3O

CAS RN 821-55-6

IUPAC Name: nonan-2-one
Synonyms: 2-nonanone
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: VKCYHJWLYTUGCC-UHFFFAOYSA-N
SMILES: CCCCCCCC(C)=O
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)