accessibility menu, dialog, popup

UserName

H3COOOOCH3

CAS RN 83-13-6

IUPAC Name: 1,3-diethyl 2-phenylpropanedioate
Synonyms: diethyl phenylmalonate
Molecular Weight (g/mol): 236.27
Molecular Formula: C13H16O4
InChi Key: FGYDHYCFHBSNPE-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C(=O)OCC)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (5)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (5)