accessibility menu, dialog, popup

UserName

NOH

CAS RN 83140-94-7

IUPAC Name: [3-(1H-pyrrol-1-yl)phenyl]methanol
Synonyms: [3-(pyrrol-1-yl)phenyl]methanol
Molecular Weight (g/mol): 173.22
Molecular Formula: C11H11NO
InChi Key: QQXDXYAGEXWXQU-UHFFFAOYSA-N
SMILES: OCC1=CC(=CC=C1)N1C=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results