accessibility menu, dialog, popup

UserName

SNNCl

CAS RN 83726-75-4

IUPAC Name: 2-chloro-4-(thiophen-2-yl)pyrimidine
Synonyms: 2-chloro-4-(thiophen-2-yl)pyrimidine
Molecular Weight (g/mol): 196.65
Molecular Formula: C8H5ClN2S
InChi Key: DTDHCBGOLGIIOA-UHFFFAOYSA-N
SMILES: ClC1=NC=CC(=N1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results