accessibility menu, dialog, popup

UserName

HOPOOHHO

CAS RN 83905-98-0

IUPAC Name: (11-hydroxyundecyl)phosphonic acid
Synonyms: 11-hydroxyundecylphosphonic acid
Molecular Weight (g/mol): 252.29
Molecular Formula: C11H25O4P
InChi Key: PPCDEFQVKBXBPS-UHFFFAOYSA-N
SMILES: OCCCCCCCCCCCP(O)(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results