accessibility menu, dialog, popup

UserName

OONH2

CAS RN 83949-32-0

IUPAC Name: 4-phenylpiperidin-1-ium-4-carboxylate
Synonyms: 4-phenylpiperidin-1-ium-4-carboxylate
Molecular Weight (g/mol): 205.26
Molecular Formula: C12H15NO2
InChi Key: DZZGGKPKWGPNJA-UHFFFAOYSA-N
SMILES: [O-]C(=O)C1(CC[NH2+]CC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results